Assay Details
| Gene Symbol: ISLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653784.1 | XP_023509552.1 | 2725 | 57 | ||
| XM_023653787.1 | XP_023509555.1 | 2732 | 57 | |||
| XM_023653790.1 | XP_023509558.1 | 3630 | 57 | |||
| XM_023653792.1 | XP_023509560.1 | 2729 | 57 | |||
| XM_023653801.1 | XP_023509569.1 | 3436 | 57 | |||
| XM_023653809.1 | XP_023509577.1 | 2398 | 57 | |||
| XM_023653820.1 | XP_023509588.1 | 2453 | 57 | |||
| XM_023653826.1 | XP_023509594.1 | 3453 | 57 | |||
| XM_023653829.1 | XP_023509597.1 | 2451 | 57 | |||
More Information
Set Membership: |
|

ISLR2
Genomic Map