Assay Details
| Gene Symbol: FSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494610.5 | XP_001494660.3 | 260 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001494610.5 transcripts.
Set Membership: |
|

FSCB
Genomic Map