Assay Details
| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650839.1 | XP_023506607.1 | 198 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023650839.1 transcripts.
Set Membership: |
|

POU3F2
Genomic Map