Assay Details
| Gene Symbol: SSTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498230.6 | XP_001498280.1 | 371 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498230.6 transcripts.
Set Membership: |
|

SSTR2
Genomic Map