Assay Details
| Gene Symbol: LOC102148661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623219.1 | XP_023478987.1 | 906 | 87 | ||
| XM_023623220.1 | XP_023478988.1 | 918 | 87 | |||
| XM_023623221.1 | XP_023478989.1 | 711 | 87 | |||
| XM_023623222.1 | XP_023478990.1 | 699 | 87 | |||
More Information
Set Membership: |
|

LOC102148661
Genomic Map