Assay Details
| Gene Symbol: MR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609762.3 | XP_005609819.1 | 172 | 83 | ||
| XM_005609764.3 | XP_005609821.1 | 174 | 83 | |||
| XM_014739668.2 | XP_014595154.1 | 175 | 83 | |||
More Information
Set Membership: |
|

MR1
Genomic Map