Assay Details
| Gene Symbol: MR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609762.3 | XP_005609819.1 | 1089 | 86 | ||
| XM_005609764.3 | XP_005609821.1 | 815 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609762.3 | XP_005609819.1 | 1089 | 86 | ||
| XM_005609764.3 | XP_005609821.1 | 815 | 86 | |||
Set Membership: |
|