Assay Details
| Gene Symbol: C5H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490773.5 | XP_001490823.1 | 315 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H1orf21| Gene Symbol: C5H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490773.5 | XP_001490823.1 | 315 | 113 | ||
Set Membership: |
|