Assay Details
| Gene Symbol: LOC106781645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014730614.2 | XP_014586100.1 | 155 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014730614.2 transcripts.
Set Membership: |
|

LOC106781645
Genomic Map