Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023620597.1 | XP_023476365.1 | 3141 | 109 | ||
| XM_023620598.1 | XP_023476366.1 | 3141 | 109 | |||
| XM_023620599.1 | XP_023476367.1 | 3251 | 109 | |||
| XM_023620600.1 | XP_023476368.1 | 3138 | 109 | |||
| XM_023620601.1 | XP_023476369.1 | 2923 | 109 | |||
| XM_023620602.1 | XP_023476370.1 | 2923 | 109 | |||
| XM_023620603.1 | XP_023476371.1 | 2992 | 109 | |||
| XM_023620604.1 | XP_023476372.1 | 2492 | 109 | |||
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map