Assay Details
| Gene Symbol: MAML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502411.4 | XP_001502461.2 | 1373 | 83 | ||
| XM_005607833.3 | XP_005607890.2 | 1372 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAML3| Gene Symbol: MAML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502411.4 | XP_001502461.2 | 1373 | 83 | ||
| XM_005607833.3 | XP_005607890.2 | 1372 | 83 | |||
Set Membership: |
|