Assay Details
| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604638.3 | XP_005604695.1 | 614 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005604638.3 transcripts.
Set Membership: |
|

GTSF1L
Genomic Map