Assay Details
| Gene Symbol: KCNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604650.3 | XP_005604707.2 | 2581 | 89 | ||
| XM_023626623.1 | XP_023482391.1 | 2117 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNS1| Gene Symbol: KCNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604650.3 | XP_005604707.2 | 2581 | 89 | ||
| XM_023626623.1 | XP_023482391.1 | 2117 | 89 | |||
Set Membership: |
|