Assay Details
| Gene Symbol: LOC100058890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003364240.3 | XP_003364288.2 | 1919 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003364240.3 transcripts.
Set Membership: |
|

LOC100058890
Genomic Map