Assay Details
| Gene Symbol: KCNJ5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741388.2 | XP_014596874.1 | 359 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014741388.2 transcripts.
Set Membership: |
|

KCNJ5
Genomic Map