Assay Details
| Gene Symbol: LOC100630879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643016.1 | XP_023498784.1 | 2345 | 90 | ||
| XM_023643031.1 | XP_023498799.1 | 2493 | 90 | |||
More Information
Set Membership: |
|

LOC100630879
Genomic Map