Assay Details
| Gene Symbol: LOC100057298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640789.1 | XP_023496557.1 | 3517 | 76 | ||
| XM_023640790.1 | XP_023496558.1 | 3340 | 76 | |||
| XM_023640791.1 | XP_023496559.1 | 2386 | 76 | |||
| XM_023640792.1 | XP_023496560.1 | 3517 | 76 | |||
More Information
Set Membership: |
|

LOC100057298
Genomic Map