Assay Details
| Gene Symbol: LOC100057298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640789.1 | XP_023496557.1 | 4572 | 91 | ||
| XM_023640790.1 | XP_023496558.1 | 4395 | 91 | |||
| XM_023640791.1 | XP_023496559.1 | 3441 | 91 | |||
| GenBank mRNA | JL621511.1 | - | 1 - 2 | 742 | 91 | |
More Information
Set Membership: |
|

LOC100057298
Genomic Map