Assay Details
Gene Symbol: LOC100058633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640819.1 | XP_023496587.1 | 299 | 103 | ||
XM_023640822.1 | XP_023496590.1 | 273 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023640822.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
