Assay Details
Gene Symbol: AIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739800.2 | XP_014595286.1 | 1490 | 95 | ||
XM_023640920.1 | XP_023496688.1 | 1490 | 95 | |||
XM_023640921.1 | XP_023496689.1 | 1386 | 95 | |||
XM_023640922.1 | XP_023496690.1 | 1453 | 95 | |||
XM_023640923.1 | XP_023496691.1 | 1284 | 95 | |||
XM_023640925.1 | XP_023496693.1 | 1934 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
