Assay Details
| Gene Symbol: PABPC1L2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633536.1 | XP_023489304.1 | 803 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PABPC1L2A| Gene Symbol: PABPC1L2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633536.1 | XP_023489304.1 | 803 | 85 | ||
Set Membership: |
|