Assay Details
| Gene Symbol: PPP1R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502693.6 | XP_001502743.3 | 159 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502693.6 transcripts.
Set Membership: |
|

PPP1R3C
Genomic Map