Assay Details
| Gene Symbol: DEPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602633.2 | XP_005602690.1 | 2304 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005602633.2 transcripts.
Set Membership: |
|

DEPP1
Genomic Map