Assay Details
| Gene Symbol: RNASE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602591.3 | XP_005602648.1 | 218 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005602591.3 transcripts.
Set Membership: |
|

RNASE4
Genomic Map