Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650078.1 | XP_023505846.1 | 255 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023650078.1 transcripts.
Set Membership: |
|

NTF4
Genomic Map