Assay Details
Gene Symbol: ZNF581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596559.3 | XP_005596616.1 | 160 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596559.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
