Assay Details
| Gene Symbol: BEND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503954.6 | XP_001504004.1 | 5828 | 88 | ||
| XM_023650873.1 | XP_023506641.1 | 537 | 88 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BEND3| Gene Symbol: BEND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503954.6 | XP_001504004.1 | 5828 | 88 | ||
| XM_023650873.1 | XP_023506641.1 | 537 | 88 | |||
Set Membership: |
|