Assay Details
| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917657.5 | XP_001917692.3 | 101 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001917657.5 transcripts.
Set Membership: |
|

CCR10
Genomic Map