Assay Details
| Gene Symbol: LRRC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493827.6 | XP_001493877.1 | 257 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493827.6 transcripts.
Set Membership: |
|

LRRC3B
Genomic Map