Assay Details
| Gene Symbol: SPSB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494656.5 | XP_001494706.3 | 2888 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPSB4| Gene Symbol: SPSB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494656.5 | XP_001494706.3 | 2888 | 80 | ||
Set Membership: |
|