Assay Details
| Gene Symbol: GPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635258.1 | XP_023491026.1 | 427 | 92 | ||
| XM_023635262.1 | XP_023491030.1 | 402 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023635258.1, XM_023635262.1 transcripts.
Set Membership: |
|

GPR3
Genomic Map