Assay Details
| Gene Symbol: LOC100059845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627826.1 | XP_023483594.1 | 1815 | 72 | ||
| XM_023627827.1 | XP_023483595.1 | 1815 | 72 | |||
| XM_023627828.1 | XP_023483596.1 | 1815 | 72 | |||
| XM_023627829.1 | XP_023483597.1 | 1815 | 72 | |||
| XM_023627830.1 | XP_023483598.1 | 1632 | 72 | |||
More Information
Set Membership: |
|

LOC100059845
Genomic Map