Assay Details
| Gene Symbol: GPR65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605386.3 | XP_005605443.1 | 102 | 145 | ||
| GenBank mRNA | JL616182.1 | - | 1 - 2 | 82 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005605386.3 transcripts.
Set Membership: |
|

GPR65
Genomic Map