Assay Details
| Gene Symbol: GPR68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605401.3 | XP_005605458.1 | 698 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005605401.3 transcripts.
Set Membership: |
|

GPR68
Genomic Map