Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736838.2 | XP_014592324.2 | 2640 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014736838.2 transcripts.
Set Membership: |
|

MGAT3
Genomic Map