Assay Details
| Gene Symbol: CHST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014738505.2 | XP_014593991.1 | 156 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014738505.2 transcripts.
Set Membership: |
|

CHST4
Genomic Map