Assay Details
| Gene Symbol: TIGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608623.3 | XP_005608680.2 | 320 | 94 | ||
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the XM_005608623.3 transcripts.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length between 86 and 100  3' Most  Equus caballus | 
Panther Classification:
 Back To Top
 Back To Top
 TIGD2
TIGD2 Genomic Map
Genomic Map Back To Top
 Back To Top