Assay Details
| Gene Symbol: LOC100056786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496636.4 | XP_001496686.2 | 410 | 107 | ||
| XM_014739810.2 | XP_014595296.1 | 406 | 107 | |||
More Information
Set Membership: |
|

LOC100056786
Genomic Map