Assay Details
| Gene Symbol: LOC100056786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496636.4 | XP_001496686.2 | 579 | 95 | ||
| XM_014739810.2 | XP_014595296.1 | 575 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100056786| Gene Symbol: LOC100056786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496636.4 | XP_001496686.2 | 579 | 95 | ||
| XM_014739810.2 | XP_014595296.1 | 575 | 95 | |||