Assay Details
| Gene Symbol: RPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610092.3 | XP_005610149.1 | 248 | 95 | ||
| XM_023641109.1 | XP_023496877.1 | 304 | 95 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023641109.1 transcripts.
Set Membership: |
|

RPS27
Genomic Map