Assay Details
| Gene Symbol: B3GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915824.4 | XP_001915859.1 | 574 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001915824.4 transcripts.
Set Membership: |
|

B3GALT2
Genomic Map