Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609321.3 | XP_005609378.1 | 1056 | 75 | ||
| XM_014739193.2 | XP_014594679.1 | 1080 | 75 | |||
| XM_014739194.2 | XP_014594680.1 | 1053 | 75 | |||
| XM_023639541.1 | XP_023495309.1 | 1084 | 75 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005609321.3, XM_014739193.2, XM_014739194.2, XM_023639541.1 transcripts.
Set Membership: |
|

GPR85
Genomic Map