Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646850.1 | XP_023502618.1 | 286 | 93 | ||
| XM_023646852.1 | XP_023502620.1 | 277 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAL3ST1| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646850.1 | XP_023502618.1 | 286 | 93 | ||
| XM_023646852.1 | XP_023502620.1 | 277 | 93 | |||
Set Membership: |
|