Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646866.1 | XP_023502634.1 | 1020 | 102 | ||
| XM_023646867.1 | XP_023502635.1 | 575 | 102 | |||
| XM_023646868.1 | XP_023502636.1 | 489 | 102 | |||
| XM_023646869.1 | XP_023502637.1 | 426 | 102 | |||
| XM_023646870.1 | XP_023502638.1 | 1552 | 102 | |||
More Information
Set Membership: |
|

LIF
Genomic Map