Assay Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619835.1 | XP_023475603.1 | 1233 | 83 | ||
| XM_023619836.1 | XP_023475604.1 | 1119 | 83 | |||
| XM_023619837.1 | XP_023475605.1 | 1121 | 83 | |||
More Information
Set Membership: |
|

KLF15
Genomic Map