Assay Details
| Gene Symbol: LOC100146643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005613732.2 | XP_005613789.2 | 102 | 79 | ||
| GenBank mRNA | KC549710.1 | - | 1 - 2 | 109 | 79 | |
| KF748746.1 | - | 1 - 2 | 47 | 79 | ||
More Information
Set Membership: |
|

LOC100146643
Genomic Map