Assay Details
| Gene Symbol: C1GALT1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501102.6 | XP_001501152.1 | 144 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501102.6 transcripts.
Set Membership: |
|

C1GALT1C1
Genomic Map