Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501347.4 | XP_001501397.1 | 434 | 103 | ||
| XM_005602325.3 | XP_005602382.1 | 773 | 103 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AVPI1| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501347.4 | XP_001501397.1 | 434 | 103 | ||
| XM_005602325.3 | XP_005602382.1 | 773 | 103 | |||
Set Membership: |
|