Assay Details
| Gene Symbol: LOC100062243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503075.3 | XP_001503125.3 | 301 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001503075.3 transcripts.
Set Membership: |
|

LOC100062243
Genomic Map