Assay Details
| Gene Symbol: LOC100062243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503075.3 | XP_001503125.3 | 536 | 76 | ||
| XM_023643143.1 | XP_023498911.1 | 422 | 76 | |||
More Information
Set Membership: |
|

LOC100062243
Genomic Map